Invitrogen™ Novex IL-10 Human Ultrasensitive ELISA Kit

A lymphokine involved in immune response

Overview
Brand: Invitrogen™

Manufacturer Part Number: KHC0103

UNSPSC: 12352203

Code: 50

Additional Details:
Additional Details: Weight: 0.86100kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10622274

Quantity Price
1 £ 516.0 / Each
EU Stock 1
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description and Specification

Specification

Regulatory Status RUO - research use only
Sample Size 50μL
Precision <0.2pg/mL
Target Interleukin 10
Sample Type Plasma, Serum, Supernatants
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
For Use With (Application) Colorimetric
Quantity 192 assays
Gene ID (Entrez) 3586
Test Time 4 hrs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.