Learn More
Invitrogen™ Novex™ IEF Sample Buffer pH 3-7 (2X)
Optimized pre-mixed IEF buffers reduce variability and ensure consistent results
Brand: Invitrogen™ LC5371
17.22 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00454kg
Description
Novex IEF Sample Buffer pH 3-7 (2X) is optimized pre-mixed IEF sample buffer for using with Novex pH 3-7 IEF gels. IEF is a sensitive technique which is affected by many factors.
Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Store in refrigerator (2–8°C). | |
2 X | |
3 to 7 | |
Novex™ IEF Gels | |
Wet Ice |
Sample Loading Buffers | |
IEF | |
IEF Sample Buffer | |
Novex™ |
For Research Use Only. Not for use in diagnostic procedures.