Learn More
Invitrogen™ iBlot™ 2 Transfer Stacks, PVDF, regular size
Consumable stacks with integrated PVDF transfer membranes for dry blotting of proteins
Brand: Invitrogen™ IB24001
146.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.50000kg
Description
Each stack contains a copper coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins.
- Increased shelf life
- Reduced and compact packaging
- Complete compatibility with iBlot 2 Gel Transfer Device
PVDF Membrane Features
- 0.2μm pore size, low fluorescence
- Higher binding capacity than nitrocellulose
- Preactivated and ready for use without any pretreatment with alcohols
- Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
- Bind proteins due to hydrophobic interactions
- Protein-binding capacity: 240μ/cm2
- The regular size (13 x 8.3cm) of this stack is used for all Novex midi gels (or other midi gel types), two Novex mini gels (or other mini gel types), and E-PAGE™ 48 or 96 gels
- The mini size (8 x 8cm) is used for all Novex mini gels (or other mini gel types)
Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
13.5 cm | |
Transfer Stack | |
0.2 μm | |
Regular, 13.5 cm x 8 cm | |
10 Blots |
8 cm | |
PVDF | |
Store at room temperature. | |
iBlot | |
Less waste, sustainable packaging, sustainable disposal |
For Research Use Only. Not for use in diagnostic procedures.