Learn More
Invitrogen™ Novex™ iBlot™ 2 Nitrocellulose Transfer Stacks
Used to transfer proteins with the iBlot 2 Gel Transfer Device
£121.00 - £147.00
Specifications
Material Nitrocellulose, 0.2 μm pore size
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15296526 View Documents Invitrogen™
IB23001
Regular, 13.5 x 8 cm £147.00
Pack of 10 View alternative products
Save Up to 
»
EU Stock 295
Add to basket
15239296 View Documents Invitrogen™
IB23002
Mini, 8 x 8 cm £121.00
Pack of 10 View alternative products
Save Up to 
»
EU Stock 357
Add to basket
Description
Description
The iBlot 2 Transfer Stacks are consumable stacks with integrated nitrocellulose transfer membranes for dry blotting of proteins. Each stack contains a copper coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins.
  • Increased shelf life
  • Reduced and compact packaging
  • Complete compatibility with iBlot 2 Gel Transfer Device
  • The nitrocellulose membrane (0.2μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer
  • Membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Protein binding capacity is 209μg/cm2
Transfer Stack Sizes
  • The regular size (13 x 8.3cm) of this stack is used for all Novex midi gels (or other midi gel types), two Novex mini gels (or other mini gel types), and E-PAGE™ 48 or 96 gels
  • The mini size (8 x 8cm) is used for all Novex mini gels (or other mini gel types)
Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Specifications
Nitrocellulose, 0.2 μm pore size
For Research Use Only. Not for use in diagnostic procedures.