Learn More
Invitrogen™ Novex™ Human Ultrasensitive Buffer Reagent Kit
Designed to be used with Human Ultrasensitive Magnetic Bead kits
Brand: Invitrogen™ LHB0003M
Code : NEW
Additional Details : Weight : 0.50000kg
Includes: Wash solution, standard reconstitution buffer, assay diluents, incubation buffer, biotin diluents, streptavidin-RPE concentrate and diluents, one 96-well flat bottom plate and black lid cover
Specifications
Novex™ Human Ultrasensitive Buffer Reagent Kit | |
Human Ultrasensitive Magnetic Bead Kits | |
Sufficient for one 96-well plate |