Learn More
Invitrogen™ Novex™ G-CSF Human ELISA Kit
Quantifies natural and recombinant human Granulocyte Colony Stimulating Factor (G-CSF) in serum, plasma, buffered solution, or cell culture medium
Brand:  Invitrogen™ KHC2031
Code : 50
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 10456283
  • £358.50 / 96 tests
Estimated Shipment
05-06-2018
Add to basket
For Research Use Only. All usage must comply with product instructions.
Specifications
Specifications
Colorimetric Detection
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers
Microplate Reader
96 assays
Plasma, Serum, Supernatants
Granulocyte colony stimulating factor 3
1440
3 hr.
G-CSF
<20pg/mL
50μL