missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Empty Gel Cassette Combs, Mini

Compatible with mini-size Empty Gel Cassettes of the same thickness (NC2010, 1.0mm; NC2015, 1.5mm)

Brand:  Invitrogen™ NC3003

Code : 50

Additional Details : Weight : 0.44000kg

 View more versions of this product

Product Code. 10338802

  • £26.89 / Pack of 25


Estimated Shipment
08-02-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Specifications

Mini-size Empty Gel Cassettes
2-well Empty Gel Cassette Comb
1.0mm
Videos