missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Bolt™ Transfer Buffer (20X)

Optimized for the transfer of proteins from Bolt Bis-Tris Plus gels to membranes for western blotting

Brand:  Invitrogen™ BT00061

Code : NEW

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 15256066

  • £143.78 / Pack of 20


Estimated Shipment
02-07-2018
Add to basket

Requires : A Bolt™ Mini Gel Tank is required for use of all Bolt gels.

Specifications

Specifications

Transfer Buffers
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Store at 4° to 25°C.
20X
1L
Videos

For Research Use Only. Not for use in diagnostic procedures.