missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ 8 to 16%, Tris-Glycine, Protein Gel, 26-well, w/adapters
Separation Range: 14 to 200 kd
Brand: Invitrogen™ WT8163A
Code : 50
Additional Details : Weight : 0.59000kg
Includes: One box of 10 gels
Description
- Provide reproducible separation of a wide range of proteins into well-resolved bands
- Individually packaged gels for optimal experiment design
- Compatible with most protein standards; Usable for native and denaturing protein assays
- Do not contain SDS and can be used to run proteins in native or in denatured form
Sample Loading Volume: 15μL; 1D Well
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
8 to 16% | |
Gradient Page | |
14 to 200 kDa | |
1 Box | |
13 cm |
Tris-Glycine | |
Protein | |
26-well, w/adapters | |
8 cm |
For Research Use Only. Not for use in diagnostic procedures.