missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NOV/CCN3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | NOV/CCN3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
NOV/CCN3 Polyclonal specifically detects NOV/CCN3 in Mouse samples. It is validated for Western Blot.Specifications
NOV/CCN3 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CCN family member 3, CCN3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog | |
NOV | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q64299 | |
4856 | |
Mouse myeloma cell line NS0-derived recombinant mouse NOV/CCN3 Ser26-Ile354 Accession # Q64299 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse NOV/CCN3 in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human NOV is observed and less than 1% cross-reactivity with recombinant mouse WISP-1 is observed. |
For Research Use Only