missing translation for 'onlineSavingsMsg'
Learn More

NOTCH3 Mouse anti-Human, APC, Clone: MHN3-21, eBioscience™

Mouse Monoclonal Antibody

£85.50 - £184.00

Specifications

Antigen Notch3
Clone MHN3-21
Target Species Human
Conjugate APC
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15578126
View Documents
Affymetrix eBioscience
17-5787-41
25 Tests £85.50
25 tests View alternative products
Save Up to 
»
Estimated Shipment
30-11-2020
Please to purchase this item. Need a web account? Register with us today!
15588126
View Documents
Affymetrix eBioscience
17-5787-42
100 Tests £184.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
30-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications Reported: This MHN3-21 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MHN3-21 antibody has been pre-titrated and tested by flow cytometric analysis of Jurkat cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. NOTCH3 is the third discovered human homologue of the Drosophilia melanogaster type I membrane protein notch. In Drosophilia, notch interaction (with its cell-bound ligands delta, serrate) establishes an intercellular signalling pathway that plays a key role in neural development. Homologues of the notch-ligands have also been identified in human. NOTCH3 functions as a receptor for membrane-bound ligands Jagged1, Jagged2, and Delta1 to regulate cell-fate determination. Mutations in NOTCH3 have been identified as the underlying cause of cerebral autosomal domit arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL).
Specifications

Specifications

Notch3
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
MHN3-21
APC
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.