missing translation for 'onlineSavingsMsg'
Learn More

NOTCH3 Mouse anti-Human, APC, Clone: MHN3-21, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5787-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15578126

  • £85.50 / 25 tests

Estimated Shipment
30-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications Reported: This MHN3-21 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MHN3-21 antibody has been pre-titrated and tested by flow cytometric analysis of Jurkat cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. NOTCH3 is the third discovered human homologue of the Drosophilia melanogaster type I membrane protein notch. In Drosophilia, notch interaction (with its cell-bound ligands delta, serrate) establishes an intercellular signalling pathway that plays a key role in neural development. Homologues of the notch-ligands have also been identified in human. NOTCH3 functions as a receptor for membrane-bound ligands Jagged1, Jagged2, and Delta1 to regulate cell-fate determination. Mutations in NOTCH3 have been identified as the underlying cause of cerebral autosomal domit arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL).
Specifications

Specifications

Notch3
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9UM47
CADASIL, CASIL
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MHN3-21
APC
NOTCH3
Liquid
NOTCH3
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
4854
Videos
Documents

For Research Use Only.