Learn More
NOTCH1 Monoclonal Antibody (22E5), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 12-5765-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: This 22E5 monoclonal antibody reacts with the extracellular domain of mouse Notch1, one of four members of the Notch family of receptors. Notch receptors are 300-kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription. Notch 1 is expressed on thymocytes, bone marrow hematopoietic stem cells, T and NK cells. Lower Notch1 expression levels can be found on B cells and monocytes. Studies show that some subsets of T-cell acute lymphoblastic leukemia (T-ALL) arise due to Notch1 chromosomal translocation with TCRbeta, which results in the expression of constitutively active Notch1. This cell surface receptor is involved in T cell lineage commitment, thymocyte development, and Th2 differentiation. Applications Reported: This 22E5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 22E5 antibody has been tested by flow cytometric analysis of mouse thymocytes.
The notch gene belongs to a family of epidermal growth factor (EGF) like homeotic genes, which encode transmembrane proteins with a variable number of cysteine-rich EGF-like repeats in the extracellular region. Four notch genes have been described in mammals: Notch1, Notch2, Notch3 and Notch4(Int-3), which have been implicated in the differentiation of the nervous system and other structures. The EGF-like proteins Delta and Serrate have been identified as ligands of Notch1. Mature Notch proteins are heterodimeric receptors derived from the cleavage of Notch pre-proteins into an extracellular subunit (NEC) containing multiple EGF-like repeats and a transmembrane subunit including intracellular region (Ntm).Specifications
NOTCH1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q01705 | |
Notch1 | |
Affinity chromatography | |
RUO | |
18128 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
22E5 | |
PE | |
Notch1 | |
9930111A19Rik; AOS5; AOVD1; Drosophila Notch homolog 1 (controlling the the ectodermal and neural cell fate in Drosophila); hN1; lin-12; major type A protein; Mis6; Motch; Motch A; mT14; N1; neurogenic locus notch homolog protein 1; NEXT; NICD; NOTCH; notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch gene homolog 1; Notch homolog 1, translocation-associated; notch receptor 1; Notch1; p300; RP23-306D20.12; RP23-306D20.12-001; Tan1; Translocation-associated notch protein TAN-1; transmembrane receptor Notch1 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.