missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Notch-2 Intracellular Domain Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | Notch-2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
Notch-2 Polyclonal specifically detects Notch-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Notch-2 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2, Notch2 | |
NOTCH2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Q04721 | |
4853 | |
E. coli-derived recombinant human Notch-2 Asp2063-Pro2413 Accession # Q04721 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Notch-2 Intracellular Domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) Notch-3 (aa 2195-2321), rhNotch-6 (aa 2251-2556) and rhNotch-4 (aa 1778-2003) is observed. |
For Research Use Only