Learn More
Integra Biosciences™ Sterile Module for Pipetboy
Use these modules with Pipetboy pipette controllers. Integra Biosciences™ Sterile Modules for Pipetboy include pipette mount, filter, filter rubber, O-ring and sterile module housing.
Brand: Integra Biosciences™ 155070
Additional Details : Weight : 0.50000kg
Description
- Complete module
- Color: Grey
- For PIPETBOY acu 2 green, red, blue and purple
Specifications
Gray | |
Nosepiece | |
Pipette mount, filter, filter rubber, O-ring and sterile module housing |