missing translation for 'onlineSavingsMsg'
Learn More

Norketamine hydrochloride, Tocris Bioscience™

Potent, non-competitive NMDA antagonist; antinociceptive. Restrictions apply. See special conditions on product page.

Brand:  Tocris (Bio-Techne) 197050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13440334

  • £389.55 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Major metabolite of ketamine that is a potent non-competitive NMDA receptor antagonist (Ki = 3.6 μM for displacement of [3H]-MK 801 in rat brain). Antinociceptive and anesthetic in vivo. R-enantiomer and S-enantiomer also available.
Specifications

Chemical Identifiers

79499-59-5
260.158
norketamine hydrochloride, 2-amino-2-2-chlorophenyl cyclohexanone hydrochloride, 2-amino-2-2-chlorophenyl cyclohexanone hcl, cyclohexanone, 2-amino-2-2-chlorophenyl-, hydrochloride, 2-amino-2-2-chlorophenyl cyclohexan-1-one;hydrochloride, +/--norketamine hydrochloride, norketamine hydrochloride 1.0 mg/ml in methanol as free base, 2-amino-2-2-chlorophenyl cyclohexan-1-one hydrochloride, cyclohexanone, 2-amino-2-2-chlorophenyl-, hydrochloride 1:1, 2-amino-2-2-chlorophenyl cyclohexan-1-one-hydrogen chloride 1/1
2-amino-2-(2-chlorophenyl)cyclohexan-1-one;hydrochloride
C12H15Cl2NO
CLPOJGPBUGCUKT-UHFFFAOYSA-N
25195026
C1CCC(C(=O)C1)(C2=CC=CC=C2Cl)N.Cl

Specifications

50mg
79499-59-5
norketamine hydrochloride, 2-amino-2-2-chlorophenyl cyclohexanone hydrochloride, 2-amino-2-2-chlorophenyl cyclohexanone hcl, cyclohexanone, 2-amino-2-2-chlorophenyl-, hydrochloride, 2-amino-2-2-chlorophenyl cyclohexan-1-one;hydrochloride, +/--norketamine hydrochloride, norketamine hydrochloride 1.0 mg/ml in methanol as free base, 2-amino-2-2-chlorophenyl cyclohexan-1-one hydrochloride, cyclohexanone, 2-amino-2-2-chlorophenyl-, hydrochloride 1:1, 2-amino-2-2-chlorophenyl cyclohexan-1-one-hydrogen chloride 1/1
C1CCC(C(=O)C1)(C2=CC=CC=C2Cl)N.Cl
260.158
260.16
Norketamine hydrochloride
C12H15Cl2NO
CLPOJGPBUGCUKT-UHFFFAOYSA-N
2-amino-2-(2-chlorophenyl)cyclohexan-1-one;hydrochloride
25195026
>99%
Videos
Documents