missing translation for 'onlineSavingsMsg'
Learn More

Norbornylene, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number p-175758
11.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
498-66-8
C7H10
94.15
MFCD00082304
JFNLZVQOOSMTJK-UHFFFAOYSA-N
bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene
10352
bicyclo[2.2.1]hept-2-ene
C1CC2CC1C=C2
This item is not returnable. View return policy
498-66-8
C7H10
94.15
MFCD00082304
JFNLZVQOOSMTJK-UHFFFAOYSA-N
bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene
10352
bicyclo[2.2.1]hept-2-ene
C1CC2CC1C=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 42.0°C to 46.0°C
Color White
Boiling Point 96.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Fieser 01,757; 02,300
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene and benzene
Stabilizer 0.1% phenolic antioxidant
Formula Weight 94.15
Percent Purity 99%
Physical Form Crystalline Mass
Chemical Name or Material Norbornylene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'