missing translation for 'onlineSavingsMsg'
Learn More

nor-Binaltorphimine dihydrochloride, Tocris Bioscience™

Selective κ antagonist

Brand:  Tocris (Bio-Techne) 0347/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13457573

  • £621.81 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A selective κ-opioid receptor antagonist.
Specifications

Chemical Identifiers

113158-34-2
734.715
nor-binaltorphimine dihydrochloride, nor-bni, nor-bni dihydrochloride, norbinaltorphimine dihydrochloride, 4,8:11,15-dimethano-20h-bisbenzofuro 2,3-a:3',2'-i dipyrido 4,3-b:3',4'-h carbazole-1,8a,10a,18-tetrol,7,12-bis cyclopropylmethyl-5,6,7,8,9,10,11,12,13,14,19a,20b-dodecahydro-, 4bs,8r,8as,10as,11r,14as,19ar,20br, d0k9he, nor-binaltorphimine dihydrochloride, solid, 17,17'-dicyclopropylmethyl-6,6',7,7'-6,6'-imino-7,7'-binorphinan-3,4',14,14'-tetrol dihydrochloride
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=C(N6)C9C12CCN(C(C1(C8)O)CC1=C2C(=C(C=C1)O)O9)CC1CC1.Cl.Cl
C40H45Cl2N3O6
JOJPJLHRMGPDPV-LZQROVCBSA-N
11957626

Specifications

nor-Binaltorphimine dihydrochloride
113158-34-2
nor-binaltorphimine dihydrochloride, nor-bni, nor-bni dihydrochloride, norbinaltorphimine dihydrochloride, 4,8:11,15-dimethano-20h-bisbenzofuro 2,3-a:3',2'-i dipyrido 4,3-b:3',4'-h carbazole-1,8a,10a,18-tetrol,7,12-bis cyclopropylmethyl-5,6,7,8,9,10,11,12,13,14,19a,20b-dodecahydro-, 4bs,8r,8as,10as,11r,14as,19ar,20br, d0k9he, nor-binaltorphimine dihydrochloride, solid, 17,17'-dicyclopropylmethyl-6,6',7,7'-6,6'-imino-7,7'-binorphinan-3,4',14,14'-tetrol dihydrochloride
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=C(N6)C9C12CCN(C(C1(C8)O)CC1=C2C(=C(C=C1)O)O9)CC1CC1.Cl.Cl
11957626
>98%
50mg
C40H45Cl2N3O6
JOJPJLHRMGPDPV-LZQROVCBSA-N
734.715
734.72
Videos
Documents