Learn More
NOP16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557693
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LRKRKVKAME VDIEERPKEL VRKPYVLNDL EAEASLPEKK GNTLSRDLID YVRYMVENHG EDYKAMAR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
NOP16 is transcriptionally regulated by c-Myc (MYC), upregulated in breast cancer, and overexpression is associated with poor patient survival (Butt et al., 2008).Specifications
NOP16 | |
Polyclonal | |
Unconjugated | |
Nop16 | |
AA409471; CGI-117; D13Wsu177e; HBV pre-S2 trans-regulated protein 3; HSPC111; HSPC185; hypothetical protein HSPC111; Nop16; NOP16 nucleolar protein; NOP16 nucleolar protein homolog; nucleolar protein 16; nucleolar protein 16 homolog; nucleolar protein 16-like; Protein ZH1; RGD1305727 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28126, 306768, 51491 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q1RP77, Q9CPT5, Q9Y3C1 | |
Nop16 | |
Recombinant protein corresponding to Human NOP16 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only