missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonyl aldehyde, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 0.8270g/mL |
Boiling Point | 93.0°C (23.0 mmHg) |
Flash Point | 63°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH3(CH2)7CHO |
Show More |
Safety and Handling
Product Identifier
- Nonyl aldehyde
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'