Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Brand: Thermo Scientific™ 919-262G
Code : 03
Additional Details : Weight : 0.25000kg
Description
- Available in low retention and extended length
- Lot-tested during production on actual workstation for which they are created
- Extended length tips compatible with 96-, 384- and 1536- deep well plates
- Low percentage coefficient of variation (%CV)
Specifications
250μL | |
Nonsterile | |
96 Tips/Rack | |
Beckman Biomek FX/NX and 3000 and Multimek AP96 Workstations |
No | |
Clear | |
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case | |
Beckman |
Technical Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.