Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Brand:  Thermo Scientific™ 917-262
UNSPSC : 41121608
Code : 03
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 10409862
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Description
Description
  • Available in low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384- and 1536- deep well plates
  • Low percentage coefficient of variation (%CV)
Specifications
Specifications
200μL
Nonsterile
96 Tips/Rack
Beckman Biomek 1000 and 2000 Workstations
No
Clear
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Beckman
Documentation Alternative Products