missing translation for 'onlineSavingsMsg'
Learn More

Nonanophenone, 97%, Thermo Scientific Chemicals

Catalog Number p-7082010
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
6008-36-2
C15H22O
218.34
MFCD00048965
PFUPABFCHVRLLY-UHFFFAOYSA-N
nonanophenone, n-nonanophenone, n-octyl phenyl ketone, 1-phenyl-1-nonanone, 1-nonanone, 1-phenyl, 1-phenyl-nonan-1-one, acmc-209mhy
80108
1-phenylnonan-1-one
CCCCCCCCC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
6008-36-2
C15H22O
218.34
MFCD00048965
PFUPABFCHVRLLY-UHFFFAOYSA-N
nonanophenone, n-nonanophenone, n-octyl phenyl ketone, 1-phenyl-1-nonanone, 1-nonanone, 1-phenyl, 1-phenyl-nonan-1-one, acmc-209mhy
80108
1-phenylnonan-1-one
CCCCCCCCC(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 14°C
Density 0.94
Boiling Point 139°C to 140°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.506
Quantity 10 g
Beilstein 2047454
Formula Weight 218.34
Percent Purity 97%
Chemical Name or Material Nonanophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'