missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Non-sterile PTFE Syringe Filter
Description
- Perfect for daily routine analysis
- Membrane is made up of PTFE material
- Low protein binding
- Hydrophilic membrane
- Offers medium chemical resistance
Specifications
PTFE | |
0.45 μm | |
For a broad range of applications | |
50/Pk. | |
25 mm |
PTFE (Hydrophilic) | |
Non-sterile | |
Polypropylene | |
Syringe Filter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction