missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Non-sterile Nylon Syringe Filter
Designed for daily routine analysis. Fisherbrand™ Non-sterile Nylon Syringe Filter features a nylon membrane in a polypropylene housing for filtering aggressive aqueous solutions from a standard syringe. 25mm diameter membranes available with 0.2 or 0.45μm pore sizes.
Brand: Fisherbrand™ 15121499
Description
- Designed for use with aggressive aqueous solutions
- Ideal for routine filtration of aqueous and solvent/aqueous mixtures
- Hydrophilic 25mm diameter nylon membrane
- Sturdy, chemically resistant polypropylene housing
Specifications
White | |
0.2 μm | |
Filtration of aggressive aqueous and solvent/aqueous mixtures | |
50/Pk. | |
25 mm |
Nylon | |
Non-sterile | |
Polypropylene | |
Syringe Filter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction