missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nomega-(p-Toluenesulfonyl)-L-arginine, 98%

Brand:  Alfa Aesar™ H63124.06

Code : NEW

Additional Details : CAS Number : 4353-32-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15469638

  • £34.45 / 5g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

Nomega-(p-Toluenesulfonyl)-L-arginine
328.39
H-Arg(Tos)-OH
4353-32-6
MFCD00077167

Specifications

Nomega-(p-Toluenesulfonyl)-L-arginine
Solid
328.39
H-Arg(Tos)-OH
4353-32-6
98%
MFCD00077167
5g
Videos
SDS