missing translation for 'onlineSavingsMsg'
Learn More

N^w-Methyl-L-arginine acetate, 99+%, Thermo Scientific™

Nitric oxide synthase inhibitor

£32.80 - £179.00

Chemical Identifiers

CAS 53308-83-1
Molecular Formula C9H20N4O4
Molecular Weight (g/mol) 248.283
MDL Number MFCD00069311
InChI Key IKPNWIGTWUZCKM-JEDNCBNOSA-N
Synonym tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
PubChem CID 135242
IUPAC Name acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
SMILES CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15403589
SDS View Documents
Thermo Scientific Alfa Aesar
J60890.MF
50mg
£179.00
50mg
View alternative products
Save Up to 
»
Estimated Shipment: 22-11-2021
Please to purchase this item. Need a web account? Register with us today!
15493579
SDS View Documents
Thermo Scientific Alfa Aesar
J60890.MA
10mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

53308-83-1
248.283
IKPNWIGTWUZCKM-JEDNCBNOSA-N
135242
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
C9H20N4O4
MFCD00069311
tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid

Specifications

Nomega-Methyl-L-arginine acetate
C9H20N4O4
6674255
tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
248.283
248.28
≥99%
50mg
53308-83-1
MFCD00069311
14,6022
IKPNWIGTWUZCKM-JEDNCBNOSA-N
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
135242
Powder
White
SDS
Product Certifications

RUO – Research Use Only