missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nomega-(2,2,4,6,7-Pentamethyl-2,3-dihydrobenzo[b]furan)-5-sulfonyl-D-arginine, 95%

Brand:  Alfa Aesar™ H63748.03

Code : NEW

Additional Details : CAS Number : 200116-81-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15465635

  • £13.78 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Nomega-(2,2,4,6,7-Pentamethyl-2,3-dihydrobenzo[b]furan)-5-sulfonyl-D-arginine
426.53
H-D-Arg(Pbf)-OH
200116-81-0
MFCD00236798

Specifications

Nomega-(2,2,4,6,7-Pentamethyl-2,3-dihydrobenzo[b]furan)-5-sulfonyl-D-arginine
Solid
426.53
H-D-Arg(Pbf)-OH
200116-81-0
95%
MFCD00236798
1g
Videos
SDS