missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Nogo Receptor/NgR Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Nogo Receptor/NgR |
---|---|
Clone | 202604 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Nogo Receptor/NgR Monoclonal specifically detects Nogo Receptor/NgR in Mouse samples. It is validated for Western Blot.Specifications
Nogo Receptor/NgR | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q99PI8 | |
65078 | |
Mouse myeloma cell line NS0-derived recombinant mouse NgR Cys27-Ser447 Accession # Q99PI8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse NgR in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human NgR. |
202604 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
NgR, NgR1, Nogo receptor, Nogo-66 receptor, NogoR, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor, RTN4R | |
RTN4R | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only