missing translation for 'onlineSavingsMsg'
Learn More

NNC 711, Tocris Bioscience™

Selective GAT-1 inhibitor

Brand:  Tocris (Bio-Techne) 177950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13439983

  • £742.35 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective inhibitor of GABA uptake by GAT-1 (IC50 values are 0.04, 171, 1700 and 622 μM for hGAT-1, rGAT-2, hGAT-3 and hBGT-1 respectively). Anticonvulsant following systemic administration in vivo. Also exhibits cognition-enhancing activity.
Specifications

Chemical Identifiers

145645-62-1
386.876
no-711 hydrochloride, 1-2-diphenylmethylene amino oxy ethyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-2-diphenylmethylene amino oxy ethyl-3-pyridinecarboxylic acid hydrochloride, 1-2-diphenylmethylene imino oxy ethyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid hydrochloride hydrochloride, 3-pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-2-diphenylmethylene amino oxy ethyl-, monohydrochloride, c21h22n2o3.hcl, no-711 hydrochloride hplc, 1-2-diphenylmethylene imino oxy ethyl-1,2,5,6-tetrahydro-3-pyridine-carboxylic acid hydrochloride, 1-2-diphenylmethylidene amino oxy ethyl-5,6-dihydro-2h-pyridine-3-carboxylic acid hydrochloride, 1-2-benzhydrylideneamino oxyethyl-3,6-dihydro-2h-pyridine-5-carboxylic acid hydrochloride
1-[2-(benzhydrylideneamino)oxyethyl]-3,6-dihydro-2H-pyridine-5-carboxylic acid;hydrochloride
C21H23ClN2O3
YZYRTEYMUTWJPL-UHFFFAOYSA-N
123738
C1CN(CC(=C1)C(=O)O)CCON=C(C2=CC=CC=C2)C3=CC=CC=C3.Cl

Specifications

50mg
145645-62-1
no-711 hydrochloride, 1-2-diphenylmethylene amino oxy ethyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-2-diphenylmethylene amino oxy ethyl-3-pyridinecarboxylic acid hydrochloride, 1-2-diphenylmethylene imino oxy ethyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid hydrochloride hydrochloride, 3-pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-2-diphenylmethylene amino oxy ethyl-, monohydrochloride, c21h22n2o3.hcl, no-711 hydrochloride hplc, 1-2-diphenylmethylene imino oxy ethyl-1,2,5,6-tetrahydro-3-pyridine-carboxylic acid hydrochloride, 1-2-diphenylmethylidene amino oxy ethyl-5,6-dihydro-2h-pyridine-3-carboxylic acid hydrochloride, 1-2-benzhydrylideneamino oxyethyl-3,6-dihydro-2h-pyridine-5-carboxylic acid hydrochloride
C1CN(CC(=C1)C(=O)O)CCON=C(C2=CC=CC=C2)C3=CC=CC=C3.Cl
386.876
386.88
NNC 711
C21H23ClN2O3
YZYRTEYMUTWJPL-UHFFFAOYSA-N
1-[2-(benzhydrylideneamino)oxyethyl]-3,6-dihydro-2H-pyridine-5-carboxylic acid;hydrochloride
123738
>99%
Videos
Documents