Learn More
NNC 63-0532, Tocris Bioscience™
Potent non-peptide NOP agonist; brain penetrant
Brand:  Tocris (Bio-Techne) 178050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13459983
  • £710.36 / 50mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Potent non-peptide agonist for the NOP receptor (EC50 = 305 nM, Ki = 7.3 nM); ∽ 12-fold selective over other receptors. Centrally active following systemic administration in vivo.
Specifications
Chemical Identifiers
250685-44-0
C27H29N3O3
8-(1-Naphthalenylmethyl)-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]decane-3-acetic acid, methyl ester
Specifications
NNC 63-0532
250685-44-0
8-(1-Naphthalenylmethyl)-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]decane-3-acetic acid, methyl ester
>98%
50mg
C27H29N3O3
443.54
Videos