missing translation for 'onlineSavingsMsg'
Learn More

NMT1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA554643

Additional Details : Weight : 0.02000kg

Product Code. 15772571

  • £426.00 / 100µL
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: MADESETAVK PPAPPLPQMM EGNGNGHEHC SDCENEEDNS YNRGGLSPAN DTGAK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

NMT1 Polyclonal antibody specifically detects NMT1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

NMT1
Polyclonal
Unconjugated
Nmt1
alternative, short form NMT-S; AW536594; Glycylpeptide N-tetradecanoyltransferase 1; long form, NMT-L; myristoyl-CoA:protein N-myristoyltransferase 1; NMT; NMT 1; Nmt1; N-myristoyltransferase 1; Peptide N-myristoyltransferase 1; type I N-myristoyltransferase
Rabbit
Antigen affinity chromatography
RUO
4836
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P30419
Nmt1
Recombinant protein corresponding to Human NMT1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only