Learn More
NMDA, Tocris Bioscience™
Selective NMDA agonist
Brand:  Tocris (Bio-Techne) 0114/500
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 500mg
 View more versions of this product
Product Code. 13447443
  • £351.33 / 500mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Prototypic NMDA receptor agonist. Also available as part of the Mixed NMDA Receptor Tocriset™ . Caged version also available (Cat. No 2224 ).
Specifications
Chemical Identifiers
6384-92-5
147.13
n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate
CHEBI:31882
CNC(CC(=O)O)C(=O)O
C5H9NO4
HOKKHZGPKSLGJE-GSVOUGTGSA-N
22880
(2R)-2-(methylamino)butanedioic acid
Specifications
NMDA
C5H9NO4
HOKKHZGPKSLGJE-GSVOUGTGSA-N
(2R)-2-(methylamino)butanedioic acid
22880
147.13
500mg
6384-92-5
n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate
CNC(CC(=O)O)C(=O)O
147.13
CHEBI:31882
>99%