Learn More
NKPD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560334
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LLTDFLCFLE IYQRRRLRVV LEVTGLDTCY PERVVGVLNA INTLLSDSHA PFIFILVVDP SILAACLESA GNMKGTADNG YLFLNRTVTL PFSVPIMGRR TKLQFLHDA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
NKPD1 Polyclonal antibody specifically detects NKPD1 in Human samples. It is validated for Immunohistochemistry.Specifications
NKPD1 | |
Polyclonal | |
Unconjugated | |
NKPD1 | |
2310015G09Rik; Nkpd1; NTPase KAP family P-loop domain containing 1; NTPase KAP family P-loop domain-containing protein 1; NTPase, KAP family P-loop domain containing 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284353 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q17RQ9 | |
NKPD1 | |
Recombinant protein corresponding to Human NKPD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only