missing translation for 'onlineSavingsMsg'
Learn More

NKH 477, Tocris Bioscience™

Water-soluble adenylyl cyclase activator

Brand:  Tocris (Bio-Techne) 160310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13449773

  • £217.56 / 10mg

Estimated Shipment date
from Supplier 24-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Water-soluble analog of forskolin (Cat. No. 1099) that is a potent activator of adenylyl cyclase; shows some selectivity for cardiac (type V) adenylyl cyclase. Stimulates bronchodilation (EC50 = 32.6 nM) and is an orally active potent positive chronotrope and hypotensive agent in vivo. Also available as part of the PKA Tocriset™ .
Specifications

Chemical Identifiers

138605-00-2
509.64
colforsin daropate, colforsin dapropate hcl, colforsin dapropate, colforsin daropate hcl, beta-alanine, n,n-dimethyl-, 3r,4ar,5s,6s,6as,10s,10ar,10bs-5-acetyloxy-3-ethenyldodecahydro-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-1h-naphtho 2,1-b pyran-6-yl ester, 3r,4ar,5s,6s,6as,10s,10ar,10bs-5-acetyloxy-3-ethenyl-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2h-benzo f chromen-6-yl 3-dimethylamino propanoate
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-5-acetyloxy-3-ethenyl-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-6-yl] 3-(dimethylamino)propanoate
C27H43NO8
RSOZZQTUMVBTMR-XGUNBQNXSA-N
444029
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)OC(=O)CCN(C)C

Specifications

NKH 477
C27H43NO8
RSOZZQTUMVBTMR-XGUNBQNXSA-N
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-5-acetyloxy-3-ethenyl-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-6-yl] 3-(dimethylamino)propanoate
444029
>98%
138605-00-2
colforsin daropate, colforsin dapropate hcl, colforsin dapropate, colforsin daropate hcl, beta-alanine, n,n-dimethyl-, 3r,4ar,5s,6s,6as,10s,10ar,10bs-5-acetyloxy-3-ethenyldodecahydro-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-1h-naphtho 2,1-b pyran-6-yl ester, 3r,4ar,5s,6s,6as,10s,10ar,10bs-5-acetyloxy-3-ethenyl-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2h-benzo f chromen-6-yl 3-dimethylamino propanoate
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)OC(=O)CCN(C)C
509.64
546.1
10mg
Videos
Documents