missing translation for 'onlineSavingsMsg'
Learn More

NKG2A/C/E, FITC, clone: 20d5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5896-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15238289

  • £74.74 / 100µg


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Description: The 20d5 monoclonal antibody reacts with the mouse NKG2A, C, and E. NKG2 molecules belong to a C-type lectin-like family of cell surface receptors expressed by mouse NK and NKT cell lineages. NKG2 molecules form heterodimeric complexes with CD94 and are responsible for recognition of non-classical MHC class I antigen Qa-1.When co-staining with NKG2A^B6 clone 16a11, it is important to stain for NKG2A-B6 first then subsequently with NKG2A/C/E, as steric hindrance has been observed if co-stained concurrently.Applications Reported: 20d5 has been reported for use in flow cytometric analysis.Applications Tested: The 20d5 antibody has been tested by flow cytometric analysis of mouse splenocytes and can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

NKG2A/C/E
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Conjugated
Klrc1, Klrc2, Klrc3
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16641, 16642, 58179
Flow Cytometry
20d5
FITC
Klrc2
CD159A, NKG2, NKG2A, C-lectin type II protein, CD159 antigen-like family member A, NK cell receptor A, NKG2-1/B activating NK receptor, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, natural killer cell l, KLRC1, cd159e, cd159c
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Videos

For Research Use Only.