missing translation for 'onlineSavingsMsg'
Learn More

NKG2A/C/E, FITC, clone: 20d5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5896-63

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15228289

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The 20d5 monoclonal antibody reacts with the mouse NKG2A, C, and E. NKG2 molecules belong to a C-type lectin-like family of cell surface receptors expressed by mouse NK and NKT cell lineages. NKG2 molecules form heterodimeric complexes with CD94 and are responsible for recognition of non-classical MHC class I antigen Qa-1.

When co-staining with NKG2AB6 clone 16a11, it is important to stain for NKG2A-B6 first then subsequently with NKG2A/C/E, as steric hindrance has been observed if co-stained concurrently.
Specifications

Specifications

NKG2A/C/E
20d5
FITC
Conjugated
Rat
10μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
NKG2A, NKG2C, NKG2E
IgG2a, kappa
RUO
Primary
Murine
Videos

For Research Use Only.