missing translation for 'onlineSavingsMsg'
Learn More

NK1.1, eVolve 655, clone: PK136, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 86-5941-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15590837

  • £74.49 / 25 tests


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The PK136 monoclonal antibody reacts with mouse NK1.1, an antigen expressed by natural killer cells and a subset of T cells in the NK1.1 mouse strains including C57BL and NZB. Several commonly used laboratory mouse strains such as BALB/c, SJL, AKR, CBA, C3H and A do not express the NK1.1 antigen. For detection of NK cells in these strains the monoclonal antibody DX5 (Cat. No. 14-5971) should be used. Simultaneous staining of C57BL/6 spleen cells with PK136 and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.Applications Reported: This PK136 antibody has been reported for use in flow cytometric analysis.Applications Tested: This PK136 antibody has been pre-titrated and tested by flow cytometric analysis of mouse splenocytes. This can be used at 5 μL per test. A test is defined as the amount of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.Laser/Filter Recommendation: eVolve™ 655 can be optimally excited with laser lines from 325-405 nm, and emits near 655 nm. We recommend using the violet (405 nm) laser line for excitation and a 630LP with a 660/40 bandpass filter, or equivalent, for detection.Fixation Recommendation: Antibodies conjugated to eVolve™ are compatible with eBioscience's fixation and permeabilization buffers.Excitation: 350-635 nm; Emission: 657 nm; Laser: Ultraviolet Laser, Violet LaserStorage and handling: For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.
Specifications

Specifications

NK1.1
PK136
eVolve 655
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Monoclonal
Flow Cytometry
5μL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CD161, NKR-P1C, Ly-55
IgG2a
RUO
Primary
Murine
Videos