missing translation for 'onlineSavingsMsg'
Learn More

NK1.1, Biotin, clone: PK136, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5941-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15239939

  • £49.20 / 50µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The PK136 monoclonal antibody reacts with mouse NK1.1, an antigen expressed by natural killer cells and a subset of T cells in the NK1.1 mouse strains including C57BL and NZB. Several commonly used laboratory mouse strains such as BALB/c, SJL, AKR, CBA, C3H and A do not express the NK1.1 antigen. For detection of NK cells in these strains the monoclonal antibody DX5 (Cat. No. 14-5971) should be used. Simultaneous staining of C57BL/6 spleen cells with PK136 and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.Applications Reported: PK136 has been reported for use in flow cytometric analysis.Applications Tested: The PK136 antibody has been tested by flow cytometric analysis of C57Bl/6 mouse splenocytes and can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

NK1.1
PK136
Biotin
Conjugated
Murine
50μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CD161, NKR-P1C, Ly-55
IgG2a
RUO
Primary
Murine
Videos