missing translation for 'onlineSavingsMsg'
Learn More

NK1.1 Monoclonal Antibody (PK136), Alexa Fluor™ 700, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£71.50 - £171.00

Specifications

Antigen NK1.1
Clone PK136
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15530367
View Documents Promotion Details
Affymetrix eBioscience
56-5941-80
25 μg
£71.50
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15540367
View Documents Promotion Details
Affymetrix eBioscience
56-5941-82
100 μg
£171.00
100µg
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The PK136 monoclonal antibody reacts with mouse NK1.1, an antigen expressed by natural killer cells and a subset of T cells in the NK1.1 mouse strains including C57BL and NZB. Several commonly used laboratory mouse strains such as BALB/c, SJL, AKR, CBA, C3H and A do not express the NK1.1 antigen. For detection of NK cells in these strains the monoclonal antibody DX5 (Cat. No. 14-5971) should be used. Simultaneous staining of C57BL/6 spleen cells with PK136 and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells. Applications Reported: This PK136 antibody has been reported for use in flow cytometric analysis. Applications Tested: This PK136 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Alexa Fluor™ 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome.

NK1.1 Monoclonal antibody specifically detects NK1.1 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

NK1.1
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
AI462337; CD161; CD161 antigen-like family member B; CD161 antigen-like family member C; CD161b; CD161c; EMBL:AAQ11375.1}; Inhibitory receptor NKR-P1B; killer cell lectin-like receptor subfamily A member 1C; killer cell lectin-like receptor subfamily B member 1B; killer cell lectin-like receptor subfamily B member 1B allele A; Killer cell lectin-like receptor subfamily B member 1B allele B; killer cell lectin-like receptor subfamily B member 1B allele C; killer cell lectin-like receptor subfamily B member 1C; killer cell lectin-like receptor subfamily B member 1D; killer cell lectin-like receptor subfamily B member 1F; KLRB1; Klrb1b; klrb1b {ECO:0000312; Klrb1c; Klrb1d; Klrb1f; klrb1f {ECO:0000250; Ly55; Ly-55; Ly55b; ly-55b; Ly55c; ly-55c; Ly55d; Ly-55d; Ly59; Ly-59; lymphocyte antigen 55 complex, locus B; lymphocyte antigen 55 complex, locus C; lymphocyte antigen 55 complex, locus D; lymphocyte antigen 55b; lymphocyte antigen 55c; Lymphocyte antigen 55d; lymphocyte antigen 59; MGC163
Klrb1b, Klrb1c
Primary
4° C, store in dark, DO NOT FREEZE!
PK136
Flow Cytometry
Alexa Fluor 700
Mouse
Mouse
P27812, P27814
17059, 80782
IgG2a κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.