missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitromethane, Technical, SLR, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Description
Nitromethane, Technical, SLR
Specifications
Melting Point | -29°C |
Color | Colorless |
pH | 6.4 |
Boiling Point | 100°C |
Packaging | Plastic Coated Glass bottle |
Quantity | 100 mL |
Formula Weight | 61.04g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Nitromethane
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'