missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane, 99+%, for analysis, Thermo Scientific Chemicals

Catalog Number p-2691938
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
75-52-5
CH3NO2
61.04
MFCD00007400
LYGJENNIWJXYER-UHFFFAOYSA-N
methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
6375
CHEBI:77701
nitromethane
C[N+]([O-])=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
75-52-5
CH3NO2
61.04
MFCD00007400
LYGJENNIWJXYER-UHFFFAOYSA-N
methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
6375
CHEBI:77701
nitromethane
C[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -29.0°C
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 100.0°C to 102.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3NO2
Refractive Index 1.3805 to 1.3825
Quantity 100 mL
Beilstein 01, 74
Residue after Evaporation 0.005% max.
Fieser 01,739; 04,357; 09,323; 13,199
Merck Index 15, 6698
Specific Gravity 1.12
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether and dimethylformamide
Viscosity 0.61 mPa.s (25°C)
Formula Weight 61.04
Percent Purity 99+%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Nitromethane, p.a.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitromethane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'