Learn More
Nitrocaramiphen hydrochloride, Tocris Bioscience™
Muscarinic antagonist, M1 > M2
Brand:  Tocris (Bio-Techne) 046910
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487713
  • £128.25 / 10mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Muscarinic antagonist with 71-fold selectivity for M1 over M2.
Specifications
Chemical Identifiers
98636-73-8
370.874
2-Diethylaminoethyl 1-(4-nitrophenyl)cyclopentanecarboxylate hydrochloride
2-(diethylamino)ethyl 1-(4-nitrophenyl)cyclopentane-1-carboxylate;hydrochloride
C18H27ClN2O4
XWQWACGTGFICFO-UHFFFAOYSA-N
25102587
CCN(CC)CCOC(=O)C1(CCCC1)C2=CC=C(C=C2)[N+](=O)[O-].Cl
Specifications
Nitrocaramiphen hydrochloride
98636-73-8
C18H27ClN2O4
XWQWACGTGFICFO-UHFFFAOYSA-N
2-(diethylamino)ethyl 1-(4-nitrophenyl)cyclopentane-1-carboxylate;hydrochloride
25102587
10mg
>98%
2-Diethylaminoethyl 1-(4-nitrophenyl)cyclopentanecarboxylate hydrochloride
CCN(CC)CCOC(=O)C1(CCCC1)C2=CC=C(C=C2)[N+](=O)[O-].Cl
370.874
370.88
Videos