missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, Reagent ACS, +99%, ACROS Organics™

Brand:  Acros Organics 415780025

Code : 29

Additional Details : CAS Number : 98-95-3 Weight : 4.51250kg Transport : UN number : 1662 Chem class : 1 Pack group : II

Quantity 2.5L
Packaging Glass bottle

 View more versions of this product

Product Code. 10529803

  • £89.80 / 2.50 L


EU Stock 7
Add to basket

Specifications

Chemical Identifiers

Nitrobenzene
99.0
ACS Reagent Grade
99+%
C6H5NO2
MFCD00007043
98-95-3
100.0
≥99%
C6H5NO2
123.11

Specifications

Nitrobenzene
88°C
210.0° to 211.0°C
5.0° to 6.0°C
98-95-3
5ppm max.
Authentic
Glass bottle
≥99%
C6H5NO2
1.5503 to 1.5523
MFCD00007043
05, 233
15, 6674
1.2050g/mL
1.205
Clear yellow
2.5L
99.0
100.0
ACS Reagent Grade
Liquid
99+%
C6H5NO2
123.11
0.005% max.
02,295; 07,251; 08,358; 17,16
0.0005 meq/g max.
Videos
SDS