missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 99%, Reagent Grade, Honeywell™

Catalog Number 15637980
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Stainless steel can
This item is not returnable. View return policy
98-95-3
C6H5NO2
123.111
MFCD00007043
LQNUZADURLCDLV-UHFFFAOYSA-N
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
7416
CHEBI:27798
nitrobenzene
C1=CC=C(C=C1)[N+](=O)[O-]
This item is not returnable. View return policy
98-95-3
C6H5NO2
123.111
MFCD00007043
LQNUZADURLCDLV-UHFFFAOYSA-N
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene
7416
CHEBI:27798
nitrobenzene
C1=CC=C(C=C1)[N+](=O)[O-]

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point 5°C to 6°C
Color Yellow
Density 1.196g/mL at 25°C
pH 8.0 to 8.5 (at 1.0000g/L 20.0°C)
Boiling Point 210°C to 211°C
Explosion Limit 0.4
Flash Point 88°C (190.4°F)
Ignition Point 899°F
Odor Pungent
Packaging Glass bottle
Quantity 1 L
UN Number UN1662
Beilstein 507540
Vapor Density 4.2 (vs air)
Vapor Pressure 0.15mmHg (20°C), 50mmHg (120°C)
Formula Weight 123.11g/mol
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material Nitrobenzene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'