missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 99%, Extra Pure, ACROS Organics™

Brand:  Acros Organics 128420100

Code : 29

Additional Details : CAS Number : 98-95-3 Weight : 12.10000kg Transport : UN number : 1662 Chem class : 1 Pack group : II

Quantity 10L
Packaging Plastic drum

 View more versions of this product

Product Code. 10308893

  • £178.40 / 10 L


Estimated Shipment
14-03-2018
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

Nitrobenzene
98.5
Extra Pure
99%
C6H5NO2
MFCD00007043
98-95-3
100.0
99%
C6H5NO2
123.11

Specifications

Nitrobenzene
88°C
210.0° to 211.0°C
5.0° to 6.0°C
98-95-3
100.0
Extra Pure
Liquid
99%
C6H5NO2
123.11
05, 233
15, 6674
1.2050g/mL
1.205
Clear yellow
10L
98.5
Authentic
Plastic drum
99%
C6H5NO2
1.5503 to 1.5523
MFCD00007043
02,295; 07,251; 08,358; 17,16
Videos
SDS