missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 99.5%, Acros Organics

Brand:  Acros Organics 447650010

Code : 29

Additional Details : CAS Number : 98-95-3 Weight : 1.95000kg Transport : UN number : 1662 Chem class : 1 Pack group : II

Quantity 1L
Packaging Glass bottle

 View more versions of this product

Product Code. 11904581

  • £39.40 / 1 L


Estimated Shipment
22-03-2018
Add to basket

Specifications

Chemical Identifiers

Nitrobenzene
For analysis
C6H5NO2
98-95-3
99.5%
123.11

Specifications

Nitrobenzene
88°C
210.0° to 211.0°C
1L
For analysis
99.5%
C6H5NO2
1.5503 to 1.5523
1.2050g/mL
1.205
5.0° to 6.0°C
98-95-3
Glass bottle
99.5%
C6H5NO2
123.11
Videos
SDS