Learn More
Palintest™ Nitrite Reagent Refill
Brand: Palintest™ AP109
Code : 04
Additional Details : CAS Number : 10043-35-3 Weight : 0.25000kg Transport : UN number : 2509 Chem class : 8 Pack group : II
Specifications
0.5ppm | |
250/Pk. | |
250/Pk. |
Safety and Handling
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Reproductive toxicity Category 1B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P264-Wash contaminated skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : boric acid, potassium hydrogensulphate,sulphanilic acid