missing translation for 'onlineSavingsMsg'
Learn More

Nitrite, Ion chromatography standard solution, Specpure™, NO2- 1000 μg/mL, Thermo Scientific Chemicals

Catalog Number p-7050957
Click to view available options
Quantity:
100 mL
500 mL
MFCD00144923
MFCD00144923

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 mL
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Nitrite Ion chromatography standard solution, NO2- 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'