missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic Acid (Certified ACS), Fisher BioReagents

Brand:  Fisher Bioreagents BP2670-500

Code : 32

Additional Details : CAS Number : 139-13-9 Weight : 0.55000kg


Quantity 500g
Packaging Poly Bottle

 View more versions of this product

Product Code. 10744534

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Nitrilotriacetic Acid
Certified ACS Grade
≥98.0%
C6H9NO6
191.14
139-13-9
Powder/Solid
≥98.0 %
N(CH2COOH)3
NTA

Specifications

Nitrilotriacetic Acid
245°C
2.3
Pass Test
Certified ACS Grade
Powder/Solid
≥98.0 %
N(CH2COOH)3
15, 6665
White
500g
139-13-9
Pass Test
Poly Bottle
≥98.0%
C6H9NO6
191.14
NTA
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Nitrilotriacetic acid

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documentation